missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Vinyl Continuous Tape

B-7569 Vinyl Continuous Tape is ideal for indoor/outdoor facility labels, including 5S labels, safety labels, warning labels, pipe markers, equipment labels and visual workplace labels.

Brand:  Brady™ 120911

Code : NEW

Additional Details : Weight : 0.91220kg

 View more versions of this product

Product Code. 15236076

  • £236.00 / 1 box
Estimated Shipment: 04-06-2024
to see stock.
Add to basket

Description

Description

  • B-7569 is ideal for indoor/outdoor facility labels, including 5S labels, safety labels, warning labels, pipe markers, equipment labels and visual workplace labels.
Specifications

Specifications

BS5609:1986 Section 2 and section 3
Permanent Acrylic, Permanent
Gloss
Thermal Transfer
Facility Identification, Floor marking, Hazard communication Identification, Pipe and valve marking, Safety Identification, Warehouse marking
5S labels, safety labels, warning labels, pipe markers, equipment labels and visual workplace labels
-20°C
1 Ea.
-4°F to +167°F
0.004 in.
30 m x 101 mm
30 m
101 mm
Continuous Vinyl Tape for BBP3X/S3XXX/i3300 Printers
Green
Vinyl
Labels
BBP30, BBP31, BBP33, BBP35, BBP37, i3300, S3000, S3100, i5300
1
1 Roll/Box
Brady Workstation, LabelMark, MarkWare
-20°C to +75°C
0.102 mm
98 ft.
3.98 in.
3.98 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers