missing translation for 'onlineSavingsMsg'
Learn More

Vinyl chloroacetate, 99%, stab. with 4-methoxyphenol

Product Code. 11377426
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377426 50 g 50g
11367426 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377426 Supplier Thermo Scientific Alfa Aesar Supplier No. L09446.18
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2549-51-1 | C4H5ClO2 | 120.532 g/mol

Vinyl chloroacetate was used to study the effect of the electron-withdrawing groups on the ligand in a series of bis(acetylacetonate)cobalt(II) derivatives. It is used for the enzyme-catalyzed, regioselective acylation of carbohydrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vinyl chloroacetate was used to study the effect of the electron-withdrawing groups on the ligand in a series of bis(acetylacetonate)cobalt(II) derivatives. It is used for the enzyme-catalyzed, regioselective acylation of carbohydrates.

Solubility
Difficult to mix in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2549-51-1
Molecular Formula C4H5ClO2
Molecular Weight (g/mol) 120.532
MDL Number MFCD00045275
InChI Key XJELOQYISYPGDX-UHFFFAOYSA-N
Synonym vinyl chloroacetate, acetic acid, chloro-, ethenyl ester, acetic acid, 2-chloro-, ethenyl ester, acetic acid, chloro-, vinyl ester, chloroacetic acid vinyl ester, vinyl chloroacetate polymer, vinyl monochloroacetate, chloroacetic acid vinyl, acetic acid, chloro-, vinyl ester, polymers, acmc-1clv7
PubChem CID 82156
IUPAC Name ethenyl 2-chloroacetate
SMILES C=COC(=O)CCl

Specifications

Density 1.19
Boiling Point 136°C
Flash Point 51°C (123°F)
Odor Pungent
Refractive Index 1.4435
Quantity 50 g
UN Number UN2589
Beilstein 1741679
Solubility Information Difficult to mix in water.
Formula Weight 120.54
Percent Purity 99%
Chemical Name or Material Vinyl chloroacetate, stabilized with 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.