missing translation for 'onlineSavingsMsg'
Learn More

Vinyl benzoate, 95%

Product Code. 11434827
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434827 100 g 100g
11444827 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11434827 Supplier Thermo Scientific Alfa Aesar Supplier No. B21805.22
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 769-78-8 | C9H8O2 | 148.161 g/mol

It is used as chemical reagent and pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as chemical reagent and pharmaceutical intermediates.

Solubility
Soluble in methanol

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 769-78-8
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00048141
InChI Key KOZCZZVUFDCZGG-UHFFFAOYSA-N
Synonym vinyl benzoate, benzoic acid vinyl ester, benzoic acid, ethenyl ester, benzoic acid, vinyl ester, unii-f1e7c1ggku, vinylester kyseliny benzoove, vinylester kyseliny benzoove czech, f1e7c1ggku, benzoic acid, ethenyl ester 9ci, benzoic acid vinyl
PubChem CID 13037
ChEBI CHEBI:84279
IUPAC Name ethenyl benzoate
SMILES C=COC(=O)C1=CC=CC=C1

Specifications

Density 1.07
Boiling Point 73°C to 74°C (8 mmHg)
Flash Point 82°C (180°F)
Refractive Index 1.529
Quantity 100 g
Beilstein 2041125
Sensitivity Moisture sensitive
Solubility Information Soluble in methanol
Formula Weight 148.16
Percent Purity 95%
Chemical Name or Material Vinyl benzoate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Vinyl benzoate
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Vinyl benzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.