missing translation for 'onlineSavingsMsg'
Learn More

Vinyl acetate, 99+%, stabilized

Product Code. 11669758 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
25mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11669758 10 L Plastic drum 10 L
10254950 25 mL Glass bottle 25mL
10113480 1 L Glass bottle 1 L
10554071 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 11669758 Supplier Thermo Scientific Acros Supplier No. 140840100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 108-05-4 | C4H6O2 | 86.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-05-4 , 123-31-9
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
InChI Key XTXRWKRVRITETP-UHFFFAOYSA-N
Synonym vinyl acetate, acetic acid ethenyl ester, acetic acid vinyl ester, ethenyl ethanoate, 1-acetoxyethylene, vinyl ethanoate, acetoxyethene, vinylacetat, vinyl acetate monomer, vinyl a monomer
PubChem CID 7904
ChEBI CHEBI:46916
IUPAC Name ethenyl acetate
SMILES CC(=O)OC=C

Specifications

Melting Point -93°C
Color Colorless
Density 0.9300g/mL
Boiling Point 72°C to 73°C
Flash Point -8°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic drum
Linear Formula CH3COOCH=CH2
Refractive Index 1.3940 to 1.396
Quantity 10 L
Beilstein 02,I,63
Fieser 01,1271; 12,565
Merck Index 15,10189
Specific Gravity 0.93
Solubility Information Solubility in water: 23g/L (20°C). Other solubilities: miscible with ethanol and ether,soluble in acetone,benzene and chloroform
Stabilizer 3 to 15ppm (hydroquinone)
Viscosity 0.43 mPa.s (20°C)
Formula Weight 86.09
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Vinyl acetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.