Learn More
Vinyl acetate, 99%, stab. with 8-12ppm hydroquinone
Description
Vinyl acetate is used in the preparation of polyvinyl acetate emulsions and polyvinyl alcohol. It acts as a precursor to polyvinyl acetate. It serves as an ingredient in furniture-glue. It is also used to prepare copolymers with other monomers such as ethylene, acrylic acid. Further, it is used to make adhesives, paints and plastics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Vinyl acetate is used in the preparation of polyvinyl acetate emulsions and polyvinyl alcohol. It acts as a precursor to polyvinyl acetate. It serves as an ingredient in furniture-glue. It is also used to prepare copolymers with other monomers such as ethylene, acrylic acid. Further, it is used to make adhesives, paints and plastics.
Solubility
Slightly miscible with water.
Notes
Light sensitive. Incompatible with acids, bases, oxidizing agents, peroxides, chlorosulfonic acid, ethylene imine, hydrochloric acid, oleum, nitric acid, sulfuric acid and 2-aminoethanol.
Specifications
Specifications
| Melting Point | °C to 100°C |
| Density | 0.932 |
| Boiling Point | 72°C to 73°C |
| Flash Point | -8°C (18°F) |
| Linear Formula | CH2=CHO2CCH3 |
| Refractive Index | 1.396 |
| Quantity | 500 mL |
| UN Number | UN1301 |
| Beilstein | 1209327 |
| Merck Index | 14,9992 |
| Show More |
Safety and Handling
- Vinyl acetate
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contain: Vinyl acetate
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.