missing translation for 'onlineSavingsMsg'
Learn More

Vinpocetine, TRC

Product Code. 30454459
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454459 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30454459 Supplier Toronto Research Chemicals Supplier No. TRCV332500100MG
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vinpocetine
CAS 42971-09-5
Molecular Formula C22 H26 N2 O2
Quantity 100 mg
Synonym Vinpocetine, Ethyl (13aS,13bS)-13a-ethyl-2,3,5,6,13a,13b-hexahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine-12-carboxylate, (3α,16α)-Eburnamenine-14-carboxylic acid ethyl ester, 1H-Indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine eburnamenine-14-carboxylic acid deriv., (+)-Apovincaminic acid ethyl ester, (+)-cis-Apovincaminic acid ethyl ester, AY 27255, Apovincaminic acid ethyl ester, Bravinton, Cavinton, Ceractin, Ethyl (+)-apovincaminate, Ethyl (+)-cis-apovincaminate, Ethyl apovincaminate, RGH 4405, TCV 3B, Ultra-Vinca, cis-Apovincaminic acid ethyl ester
InChI Formula InChI=1S/C22H26N2O2/c1-3-22-11-7-12-23-13-10-16-15-8-5-6-9-17(15)24(19(16)20(22)23)18(14-22)21(25)26-4-2/h5-6,8-9,14,20H,3-4,7,10-13H2,1-2H3/t20-,22+/m1/s1
SMILES CCOC(=O)C1=C[C@]2(CC)CCCN3CCc4c([C@H]23)n1c5ccccc45
IUPAC Name Ethyl (13aS,13bS)-13a-ethyl-2,3,5,6,13a,13b-hexahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine-12-carboxylate
Molecular Weight (g/mol) 350.45
Formula Weight 350.1994
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.