missing translation for 'onlineSavingsMsg'
Learn More

Vinpocetine, MedChemExpress

Product Code. p-200095875
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16491656

Brand: Medchem Express HY13295

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vinpocetine (Ethyl apovincaminate) is a derivative of the alkaloid Vincamine that blocks voltage-gated Na+ channels.

The IC50 value of Vinpocetine on direct IKK inhibition in the cell-free system is 17.17 μM. Vinpocetine is a phosphodiesterase (PDE) inhibitor and inhibits NF-κB-dependent inflammatory responses by directly targeting IκB kinase complex (IKK), and has been widely used for the treatment of cerebrovascular disorders.

Specifications

Chemical Name or Material Vinpocetine
CAS 42971-09-5
Color White
Purity Grade Notes Research
Molecular Formula C22H26N2O2
Quantity 500 mg
Synonym Ethyl apovincaminate
Solubility Information Ethanol : 7.14 mg/mL (20.37 mM; Need ultrasonic) ∣DMSO : 6.25 mg/mL (17.83 mM; Need ultrasonic)
SMILES CCOC(C1=C[C@]2(CC)CCCN3CCC(C4=C(C=CC=C4)N51)=C5[C@]23[H])=O
Molecular Weight (g/mol) 350.45
Formula Weight 350.45
Percent Purity 99.34%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.