missing translation for 'onlineSavingsMsg'
Learn More

Vinorelbine Tartrate, Mikromol™

Product Code. 18072200
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18072200 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18072200 Supplier Mikromol Supplier No. MM1252.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Vinorelbine Tartrate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vinorelbine Ditartrate
CAS 125317-39-7
Analyte or Component Names Vinorelbine Ditartrate
Shipping Condition Ice pack (-20°C)
Molecular Formula C45 H54 N4 O8 . 2 C4 H6 O6
Certifications/Compliance ISO 17025
Synonym Vinorelbine tartrate, Methyl (3aR,4R,5S,5aR,10bR,13aR)-4-(acetyloxy)-3a-ethyl-9-[(6R,8S)-4-ethyl-8-(methoxycarbonyl)-1,3,6,7,8,9-hexahydro-2,6-methano-2H-azacyclodecino[4,3-b]indol-8-yl]-5-hydroxy-8-methoxy-6-methyl-3a,4,5,5a,6,11,12,13a-octahydro-1H-indolizino[8,1-cd]carbazole-5-carboxylate dihydrogen bis[(2R,3R)-2,3-dihydroxybutanedioate]
InChI Formula InChI=1S/C45H54N4O8.2C4H6O6/c1-8-27-19-28-22-44(40(51)55-6,36-30(25-48(23-27)24-28)29-13-10-11-14-33(29)46-36)32-20-31-34(21-35(32)54-5)47(4)38-43(31)16-18-49-17-12-15-42(9-2,37(43)49)39(57-26(3)50)45(38,53)41(52)56-7;2*5-1(3(7)8)2(6)4(9)10/h10-15,19-21,28,37-39,46,53H,8-9,16-18,22-25H2,1-7H3;2*1-2,5-6H,(H,7,8)(H,9,10)/t28-,37-,38+,39+,42+,43+,44-,45-;2*1-,2-/m011/s1
SMILES CCC1=C[C@@H]2CN(C1)Cc3c([nH]c4ccccc34)[C@@](C2)(C(=O)OC)c5cc6c(cc5OC)N(C)[C@H]7[C@](O)([C@H](OC(=O)C)[C@]8(CC)C=CCN9CC[C@]67[C@H]89)C(=O)OC.O[C@H]([C@@H](O)C(=O)O)C(=O)O.O[C@H]([C@@H](O)C(=O)O)C(=O)O
IUPAC Name Methyl (3aR,4R,5S,5aR,10bR,13aR)-4-(acetyloxy)-3a-ethyl-9-[(6R,8S)-4-ethyl-8-(methoxycarbonyl)-1,3,6,7,8,9-hexahydro-2,6-methano-2H-azacyclodecino[4,3-b]indol-8-yl]-5-hydroxy-8-methoxy-6-methyl-3a,4,5,5a,6,11,12,13a-octahydro-1H-indolizino[8,1-cd]carbazole-5-carboxylate dihydrogen bis[(2R,3R)-2,3-dihydroxybutanedioate]
Molecular Weight (g/mol) 1079.11
Quantity 100 mg
Formula Weight 1078.427 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.