missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vinorelbine ditartrate, Tocris Bioscience™
Selective mitotic microtubule antagonist
Chemical Identifiers
| CAS | 125317-39-7 |
|---|---|
| Molecular Formula | C53H66N4O20 |
| Molecular Weight (g/mol) | 1079.119 |
| InChI Key | CILBMBUYJCWATM-HJBYVSOESA-N |
| Synonym | vinorelbine ditartrate |
| PubChem CID | 124080885 |
| SMILES | CCC1=CC2CC(C3=C(CN(C2)C1)C4=CC=CC=C4N3)(C5=C(C=C6C(=C5)C78CCN9C7C(C=CC9)(C(C(C8N6C)(C(=O)OC)O)OC(=O)C)CC)OC)C(=O)OC.C(C(C(=O)O)O)(C(=O)O)O.C(C(C(=O)O)O)(C(=O)O)O |
Description
Selective mitotic microtubule antagonist that exhibits > 20 fold selectivity over axonal microtubules. Inhibits proliferation of multiple human tumor cell lines (IC50 = 1.25 nM in HeLa cells) and blocks metaphase/anaphase transition by suppression of microtubule dynamics (IC50 = 3.8 nM). Reduces spindle length by 29% and inhibits microtubule polymerization at micromolar concentrations.Chemical Identifiers
| 125317-39-7 | |
| 1079.119 | |
| vinorelbine ditartrate | |
| CCC1=CC2CC(C3=C(CN(C2)C1)C4=CC=CC=C4N3)(C5=C(C=C6C(=C5)C78CCN9C7C(C=CC9)(C(C(C8N6C)(C(=O)OC)O)OC(=O)C)CC)OC)C(=O)OC.C(C(C(=O)O)O)(C(=O)O)O.C(C(C(=O)O)O)(C(=O)O)O |
| C53H66N4O20 | |
| CILBMBUYJCWATM-HJBYVSOESA-N | |
| 124080885 |
Specifications
| 125317-39-7 | |
| vinorelbine ditartrate | |
| CCC1=CC2CC(C3=C(CN(C2)C1)C4=CC=CC=C4N3)(C5=C(C=C6C(=C5)C78CCN9C7C(C=CC9)(C(C(C8N6C)(C(=O)OC)O)OC(=O)C)CC)OC)C(=O)OC.C(C(C(=O)O)O)(C(=O)O)O.C(C(C(=O)O)O)(C(=O)O)O | |
| 124080885 | |
| >98% |
| C53H66N4O20 | |
| CILBMBUYJCWATM-HJBYVSOESA-N | |
| 1079.119 | |
| 1079.1 | |
| Vinorelbine ditartrate |
Safety and Handling
Recommended Storage : Store at -20°C
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.