missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vinorelbine ditartrate, MedChemExpress
Description
Vinorelbine (ditartrate) is an anti-mitotic agent which inhibits the proliferation of Hela cells with IC50 of 1.25 nM.
Specifications
Specifications
| Chemical Name or Material | Vinorelbine ditartrate |
| CAS | 125317-39-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C53H66N4O20 |
| Quantity | 10 mg |
| Synonym | KW-2307 Nor-5'-anhydrovinblastine ditartrate |
| Solubility Information | DMSO : ≥ 100 mg/mL (92.67 mM) ∣H2O : ≥ 100 mg/mL (92.67 mM) |
| SMILES | CC[C@@]1(C=CCN2CC3)[C@@]2([H])[C@@]3(C(C=C([C@@](C4=C5C(C=CC=C6)=C6N4)(CC(C=C(CC)C7)([H])CN7C5)C(OC)=O)C(OC)=C8)=C8N9C)[C@]9([H])[C@](C(OC)=O)(O)[C@@H]1OC(C)=O.O=C(O)[C@H](O)[C@@H](O)C(O)=O.O=C(O)[C@H](O)[C@@H](O)C(O)=O |
| Molecular Weight (g/mol) | 1079.11 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?