missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vinorelbine-d3 Bitartrate, TRC
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Specifications
Specifications
| Chemical Name or Material | Vinorelbine-d3 Bitartrate |
| Molecular Formula | C53H63D3N4O20 |
| Quantity | 1 mg |
| Synonym | 3',4'-Didehydro-4'-deoxy-C'-norvincaleukoblastine-d3 Ditartrate, Nor-5'-anhydrovinblastine-d3 Ditartrate, Eunades-d3, Navelbine-d3 |
| InChI Formula | InChI=1S/C45H54N4O8.C4H6O6/c1-8-27-19-28-22-44(40(51)55-6,36-30(25-48(23-27)24-28)29-13-10-11-14-33(29)46-36)32-20-31-34(21-35(32)54-5)47(4)38-43(31)16-18-49-17-12-15-42(9-2,37(43)49)39(57-26(3)50)45(38,53)41(52)56-7;5-1(3(7)8)2(6)4(9)10/h10-15,19-21,28,37-39,46,53H,8-9,16-18,22-25H2,1-7H3;1-2,5-6H,(H,7,8)(H,9,10)/t28?,37-,38+,39+,42+,43+,44+,45-;/m0./s1/i6D3; |
| SMILES | [H]C(C=C(CC)C1)(C[C@]2(C(C(OC)=C3)=CC4=C3N(C)[C@@]5([H])[C@@](O)(C(OC)=O)[C@H](OC(C)=O)[C@@]6(CC)[C@@]7([H])[C@]54CCN7CC=C6)C(OC([2H])([2H])[2H])=O)C[N@]1CC8=C2NC9=C8C=CC=C9.OC(C(O)C(O)=O)C(O)=O |
| Molecular Weight (g/mol) | 1082.12 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction