missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vinculin, Mouse anti-Human, Clone: VCL/2572, Novus Biologicals™
Mouse Monoclonal Antibody
£221.00 - £395.00
Specifications
| Antigen | Vinculin |
|---|---|
| Clone | VCL/2572 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18251066
|
Novus Biologicals
NBP2-79806-20UG |
20 μg |
£221.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18205088
|
Novus Biologicals
NBP2-79806-100UG |
100 μg |
£395.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Vinculin Monoclonal specifically detects Vinculin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
| Vinculin | |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cellular Markers, Phospho Specific | |
| 10 mM PBS with 0.05% BSA | |
| CMD1W, CMH15, Metavinculin, MVCL, vinculin | |
| VCL | |
| IgG1 κ | |
| Protein A or G purified |
| VCL/2572 | |
| Western Blot 1-2 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P18206, P18206 | |
| 7414 | |
| Recombinant fragment (around aa 174-322) of human Vinculin protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title