missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Vinculin Monoclonal Antibody (C4), Invitrogen™

Mouse Monoclonal Antibody
£161.00 - £389.00
Specifications
| Antigen | Vinculin |
|---|---|
| Clone | C4 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17928602
|
Invitrogen™
MA550898 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17980633
![]() |
Invitrogen™
MA550897 |
200 μL |
£389.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Vinculin (VCL) is a cytoskeletal protein associated with cell-cell and cell-extracellular matrix adherens-type junctions. It functions as one of several interacting proteins involved in anchoring F-actin to the membrane. It has been shown that a sequence of molecular interactions might be involved in the transmembrane assembly of adhesion plaques. In the assembly of adhesion plaques, the beta subunit of integrin binds to talin. Talin binds to vinculin that interacts with alpha-actinin and possibly with itself. Since alpha-actinin binds to and cross-links actin filaments, vinculin represents a key element in the transmembrane linkage of the extracellular matrix to the cytoplasmic microfilament system. Mutations in the gene can result in cardiomyopathy dilated 1W or familial hypertrophic 15.Specifications
| Vinculin | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| 9430097D22; AA571387; AI462105; AW545629; CMD1W; CMH15; epididymis luminal protein 114; HEL114; metavinculin; meta-vinculin; metavinculin {ECO:0000250; MV; MVCL; RP11-178G16.3; UniProtKB:P18206}; unnamed protein product; Vcl; vcl {ECO:0000250; VIN; vinc; vinc 1; VINC1; Vinculin; vinculin {ECO:0000250 | |
| VCL | |
| IgG2b κ | |
| Protein A/G | |
| VCL |
| C4 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat, Canine, Rabbit, Bovine, Pig, Chicken, Guinea Pig | |
| P12003, P18206, P26234, P85972, Q64727 | |
| 100357160, 100722183, 22330, 305679, 396422, 396974, 479252, 7414, 783645 | |
| Recombinant Human/Mouse/Rat VCL protein, Lys1020-Gln1134 (Accession #P18206). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title