missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vinculin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41237-0.025mg
This item is not returnable.
View return policy
Description
Vinculin Polyclonal specifically detects Vinculin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| Vinculin | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| VCL | |
| Antibody was raised against a 16 amino acid peptide near the carboxy terminus of human Vinculin. The immunogen is located within amino acids 1060 - 1110 of Vinculin . Amino Acid Squence: LGRTNISDEESEQATE | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Vinculin antibody is human, mouse and rat specific. At least three isoforms of Vinculin are known to exist. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 1:10-1:500 | |
| CMD1W, CMH15, Metavinculin, MVCL, vinculin | |
| Rabbit | |
| 120 kDa | |
| 0.025 mg | |
| Cellular Markers, Cytoskeleton Markers | |
| 7414 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction