missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Vincristine sulfate
Specifications
Specifications
| Content And Storage | 4°C, sealed storage, away from moisture and light∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light) |
| Molecular Weight (g/mol) | 923.04 g/mol |
| Shipping Condition | Room temperature |
| For Use With (Application) | Cancer-programmed cell death |
| CAS | 2068-78-2 |
| Product Type | Natural Products |
| Purity | 97.83% |
| Solubility | DMSO : 83.33 mg/mL (90.28 mM∣ultrasonic and warming and heat to 80°C) ∣H2O : 50 mg/mL (54.17 mM∣Need ultrasonic) |
| Synonym | Leurocristine sulfate∣NSC-67574 sulfate∣22-Oxovincaleukoblastine sulfate |
| Target | Microtubule/Tubulin∣Apoptosis |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?