missing translation for 'onlineSavingsMsg'
Learn More

HC Innovations Vinco-SanWipe Cleaning & Sanitising Facilities Anti-Bac and Anti-Viral Wipe

Product Code. p-7221537
Click to view available options
Packaging Quantity:
1000 Wipes
150 Wipes
500 Wipes
This item is not returnable. View return policy

Product Code. 17234184

Brand: HC Innovations CP128

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Formulated for use in gyms, airports, workplaces, facilities management, industrial areas, showrooms and offices

  • Can be used on hands and surfaces, for best results use 1 wipe to remove soiling and the second wipe to sanitize.
  • These facilities wipes are best used on hands and hard surfaces such as desks and equipment in offices, door handles, grab handles, machinery, keyboards, washroom areas and other areas where there is high foot fall.

Specifications

Product Type Sanitizing Wipes
Autoclavable Not Autoclavable
Basis Weight 25 gsm
Color White
Content And Storage Store in a Dry Place
Disposable Single-use
Edges Angled
Dimensions (L x W) 200 x 200 mm
For Use With (Application) Cleaning and sanitizing
Packaging Quantity 150 Wipes
Ply 1-ply
Saturation Solution 14.3 Chemical / 85.7 Water
Sterility Non-sterile
Includes Fabric, Chemical
Green Features Recyclable
Care Instructions Keep upright
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.