missing translation for 'onlineSavingsMsg'
Learn More

HC Innovations Vinco-SanWipe Cleaning & Sanitising Facilities Anti-Bac and Anti-Viral Wipe

Product Code. 17264204
Change view
Click to view available options
Packaging Quantity:
1000 Wipes
150 Wipes
500 Wipes
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity
17264204 500 Wipes
17234184 150 Wipes
17254204 1000 Wipes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17264204 Supplier HC Innovations Supplier No. CP147
Stock Up and Save Up to 60%
Use promo code 28009
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Formulated for use in gyms, airports, workplaces, facilities management, industrial areas, showrooms and offices

  • Can be used on hands and surfaces, for best results use 1 wipe to remove soiling and the second wipe to sanitize.
  • These facilities wipes are best used on hands and hard surfaces such as desks and equipment in offices, door handles, grab handles, machinery, keyboards, washroom areas and other areas where there is high foot fall.

Specifications

Product Type Sanitizing Wipe
Material Polypropylene
Autoclavable Not Autoclavable
Basis Weight 25 gsm
Color White
Content And Storage Store in a Dry Place
Disposable Single-use
Edges Angled
Dimensions (L x W) 200 x 200 mm
For Use With (Application) Cleaning and sanitizing
Packaging Quantity 500 Wipes
Ply 1-ply
Saturation Solution 14.3 Chemical / 85.7 Water
Sterility Non-sterile
Includes Fabric, Chemical
Green Features Recyclable
Care Instructions Keep upright
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.