missing translation for 'onlineSavingsMsg'
Learn More

Vinburnine, MedChemExpress

Product Code. 16418666
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16418666 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16418666 Supplier Medchem Express Supplier No. HYB1180
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Vincamone is a vinca alkaloid and a metabolite of vincamine, is a vasodilator.

Vincamone is a vinca alkaloid and a metabolite of vincamine, is a vasodilator.

Specifications

Chemical Name or Material Vinburnine
CAS 4880-88-0
Color White
Purity Grade Notes Research
Molecular Formula C19H22N2O
Quantity 100 mg
Synonym (-)-Eburnamonine (-)-Vincamone
Solubility Information DMSO : 8.33 mg/mL (28.30 mM; Need ultrasonic)
SMILES O=C1C[C@@]2(CC)[C@@]3([H])C(N1C4=CC=CC=C54)=C5CCN3CCC2
Molecular Weight (g/mol) 294.39
Formula Weight 294.39
Percent Purity 99.42%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.