Learn More
Thermo Scientific™ Vinblastine Sulfate, 98%
Anticancer agent; microtubule disrupter; Induces apoptosis
£143.00 - £520.00
Chemical Identifiers
| CAS | 143-67-9 |
|---|---|
| Formula Weight | 909.06g/mol |
| Synonym | Vincaleukblastine sulfate |
| Content And Storage | 2°C to 8°C |
| Format | Powder |
| For Use With (Application) | Anticancer agent; microtubule disrupter; Induces apoptosis |
| Description | White |
| Melting Point | 267°C |
| Solubility | Soluble in water or DMSO |
| Assay | 98% |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15415799
|
Thermo Scientific™
J63598.MA |
10 mg |
£143.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15425799
|
Thermo Scientific™
J63598.MF |
£520.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 143-67-9 | |
| Vincaleukblastine sulfate | |
| Powder | |
| White | |
| Soluble in water or DMSO |
| 909.06g/mol | |
| 2°C to 8°C | |
| Anticancer agent; microtubule disrupter; Induces apoptosis | |
| 267°C | |
| 98% |
Specifications
| White | |
| Powder | |
| 143-67-9 | |
| 909.06g/mol | |
| 267°C | |
| 3659812 |
| 2°C to 8°C | |
| Anticancer agent; microtubule disrupter; Induces apoptosis | |
| C46H60N4O13S | |
| Vincaleukblastine sulfate | |
| Soluble in water or DMSO | |
| 98% |
Safety and Handling
missing translation for 'einecsNumber' : 205-606-0
Recommended Storage : Keep cold
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.