missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Vinblastine sulfate, 96.0-102.0%

Product Code. 10470861 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
Packaging:
Glass bottle
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10470861 5 mg Glass bottle 5mg
10289223 25 mg Glass bottle 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10470861 Supplier Thermo Scientific Chemicals Supplier No. 203450050
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 143-67-9 | C46H60N4O13S | 909.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-67-9
Molecular Formula C46H60N4O13S
Molecular Weight (g/mol) 909.06
MDL Number MFCD08706468,MFCD00082457,MFCD08706468
InChI Key KDQAABAKXDWYSZ-JKDPCDLQSA-N
Synonym vinblastine sulfate
PubChem CID 131709225
IUPAC Name methyl (1R,9R,10S,11R,12R,19R)-11-(acetyloxy)-12-ethyl-4-[(13S,15S,17S)-17-ethyl-17-hydroxy-13-(methoxycarbonyl)-1,11-diazatetracyclo[13.3.1.0⁴,¹².0⁵,¹⁰]nonadeca-4(12),5,7,9-tetraen-13-yl]-10-hydroxy-5-methoxy-8-methyl-8,16-diazapentacyclo[10.6.1.0¹,⁹.0²,⁷.0¹⁶,¹⁹]nonadeca-2(7),3,5,13-tetraene-10-carboxylate; sulfuric acid
SMILES OS(O)(=O)=O.CC[C@]1(O)C[C@H]2CN(C1)CCC1=C(NC3=CC=CC=C13)[C@@](C2)(C(=O)OC)C1=CC2=C(C=C1OC)N(C)[C@@H]1[C@]22CCN3CC=C[C@](CC)([C@@H]23)[C@@H](OC(C)=O)[C@]1(O)C(=O)OC

Specifications

Melting Point 267.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96.0 to 102% (HPLC)
Packaging Glass bottle
Quantity 5 mg
Merck Index 15, 10179
Specific Rotation Condition −21.70° (21°C c=2,CH3OH)
Specific Rotation -21.7
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohols,acetone,ethyl acetate,,chloroform,practically insoluble in petroleum ether
Formula Weight 909.05
Percent Purity 96.0-102%
Physical Form Crystalline Powder
Chemical Name or Material Vinblastine sulfate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Vinblastine sulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Vinblastine sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.