Learn More
Invitrogen™ Vimentin Polyclonal Antibody
Description
PA1-10003 can be used to study stem cells and generally to reveal the intermediate filament cytoskeleton. The immunogen used to generate this antibody was full length recombinant human vimentin. The antibody works well on all mammals tested to date, and it was generated in chicken by standard procedures and immunoglobulin was extracted from egg yolk.
Specifications
Specifications
| Antigen | Vimentin |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Vim |
| Gene Accession No. | P02543, P08670, P09654, P20152, P31000, P48616 |
| Gene Alias | cb28; class-III intermediate microfilament; CTRCT30; epididymis luminal protein 113; FLJ36605; Hel113; intermediate filament; intermediate filament protein; RP11-124N14.1; similar to vimentin; VIM; VIME; Vimentin; vimentin C-terminus |
| Gene Symbols | Vim |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.