missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Vimentin Monoclonal antibody specifically detects Vimentin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Vimentin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | GT11512 |
| Conjugate | Unconjugated |
| Formulation | PBS |
| Gene Alias | epididymis secretory sperm binding protein, FLJ36605, VIM, vimentin |
| Host Species | Mouse |
| Immunogen | Carrier-protein conjugated synthetic peptide corresponding to residues around human Vimentin (phospho Ser56). The exact sequence is proprietary. |
| Purification Method | Protein G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?