missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Vimentin Monoclonal Antibody (V9)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA511883

407.55 GBP valid until 2025-12-31
BEST PRICE on Promo! Use promo code "24159" to get your promotional price.


Product Code. 11344533

  • £429.00 / 500µL
Spotlight
Use promo code 24159
View all promotions

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Description

Description

MA5-11883 targets Vimentin in IHC (P) applications and shows reactivity with Bovine, Canine, Chicken, Equine, Feline, Gerbil, Hamster, Human, mouse, Non-human primate, Porcine, Rabbit, and Rat samples. The MA5-11883 immunogen is purified vimentin from pig eye lens.

Vimentin is a developmentally regulated intermediate filament protein (IFP) found in cells of mesenchymal origin. It is believed to be involved with the intracellular transport of proteins between the nucleus and plasma membrane. Unlike other IFP proteins, vimentin is expressed, along with desmin, during the early stages of cellular development. During the development process, vimentin is exchanged for new, tissue-specific IFPs. Vimentin has been implicated to be involved in the rate of steroid synthesis via its role as a storage network for steroidogenic cholesterol containing lipid droplets. Vimentin phosphorylation by a protein kinase causes the breakdown of intermediate filaments and activation of an ATP and myosin light chain dependent contractile event. This results in cytoskeletal changes that facilitate the interaction of the lipid droplets within mitochondria, and subsequent transport of cholesterol to the organelles leading to an increase in steroid synthesis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Vimentin
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P02543, P08670, P09654, P20152, P31000, P48616, P48670
Vim
Purified vimentin from pig eye lens.
500 μL
Primary
Bovine, Canine, Chicken, Horse, Feline, Gerbil, Hamster, Human, Mouse, Monkey, Pig, Pig, Rat
Antibody
IgG1 κ
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
V9
Unconjugated
Vim
cb28; class-III intermediate microfilament; CTRCT30; epididymis luminal protein 113; FLJ36605; HEL113; intermediate filament; intermediate filament protein; RP11-124N14.1; similar to vimentin; VIM; VIME; Vimentin; vimentin C-terminus
Mouse
Protein G
RUO
100008924, 100056088, 100522394, 100689099, 101091951, 22352, 280955, 420519, 477991, 7431, 81818
4°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.