Learn More
Invitrogen™ Vimentin Monoclonal Antibody (SP20)

Rabbit Monoclonal Antibody
Brand: Invitrogen™ MA516409
Description
Predicted to react with Rat, Hamster, Cow, and Xenopus laevis based on sequence homology. Recommended Postive Controls - WB: U-2 OS, Hu tonsil and HeLa cell lysates. Flow: HeLa cells. ICC/IF: HAP1-VIM cells, human limbal epithelial cells. IHC-P: Human breast cancer and melanoma tissue. IHC-Fr: Colorectal cancer tissue.
Vimentin is a developmentally regulated intermediate filament protein (IFP) found in cells of mesenchymal origin. It is believed to be involved with the intracellular transport of proteins between the nucleus and plasma membrane. Unlike other IFP proteins, vimentin is expressed, along with desmin, during the early stages of cellular development. During the development process, vimentin is exchanged for new, tissue-specific IFPs. Vimentin has been implicated to be involved in the rate of steroid synthesis via its role as a storage network for steroidogenic cholesterol containing lipid droplets. Vimentin phosphorylation by a protein kinase causes the breakdown of intermediate filaments and activation of an ATP and myosin light chain dependent contractile event. This results in cytoskeletal changes that facilitate the interaction of the lipid droplets within mitochondria, and subsequent transport of cholesterol to the organelles leading to an increase in steroid synthesis.
Specifications
| Vimentin | |
| Monoclonal | |
| 0.006 mg/mL | |
| PBS with 1% BSA and 0.1% sodium azide; pH 7.20 | |
| P08670, P20152 | |
| Vim | |
| Recombinant human vimentin protein. | |
| 500 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| SP20 | |
| Unconjugated | |
| Vim | |
| cb28; class-III intermediate microfilament; CTRCT30; epididymis luminal protein 113; FLJ36605; HEL113; intermediate filament; intermediate filament protein; RP11-124N14.1; similar to vimentin; VIM; VIME; Vimentin; vimentin C-terminus | |
| Rabbit | |
| Protein A | |
| RUO | |
| 22352, 7431 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.