missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Vimentin Monoclonal Antibody (2D1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547401
This item is not returnable.
View return policy
Description
Vimentin Monoclonal Antibody for Western Blot, ICC/IF, IHC
Vimentin is a developmentally regulated intermediate filament protein (IFP) found in cells of mesenchymal origin. It is believed to be involved with the intracellular transport of proteins between the nucleus and plasma membrane. Unlike other IFP proteins, vimentin is expressed, along with desmin, during the early stages of cellular development. During the development process, vimentin is exchanged for new, tissue-specific IFPs. Vimentin has been implicated to be involved in the rate of steroid synthesis via its role as a storage network for steroidogenic cholesterol containing lipid droplets. Vimentin phosphorylation by a protein kinase causes the breakdown of intermediate filaments and activation of an ATP and myosin light chain dependent contractile event. This results in cytoskeletal changes that facilitate the interaction of the lipid droplets within mitochondria, and subsequent transport of cholesterol to the organelles leading to an increase in steroid synthesis.
Specifications
| Vimentin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 5mM sodium azide | |
| P08670, P31000 | |
| Vim | |
| Full length recombinant human vimentin expressed in and purified from E. coli. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2a |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 2D1 | |
| Unconjugated | |
| Vim | |
| cb28; class-III intermediate microfilament; CTRCT30; epididymis luminal protein 113; FLJ36605; HEL113; intermediate filament; intermediate filament protein; RP11-124N14.1; similar to vimentin; VIM; VIME; Vimentin; vimentin C-terminus | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 7431, 81818 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction