missing translation for 'onlineSavingsMsg'
Learn More

Vimentin Antibody (VM452), PE, Novus Biologicals™

Product Code. 18406811 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406811 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406811 Supplier Novus Biologicals Supplier No. NBP233060PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Vimentin Monoclonal antibody specifically detects Vimentin in Human, Porcine, Bovine, Canine, Chicken, Feline, Goat, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Protein Array, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vimentin
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Protein Array, KnockDown
Classification Monoclonal
Clone VM452
Conjugate PE
Formulation PBS
Gene Alias epididymis secretory sperm binding protein, FLJ36605, VIM, vimentin
Host Species Mouse
Immunogen Recombinant full-length human vimentin protein (Uniprot: P08670)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 7431
Target Species Human, Pig, Bovine, Canine, Chicken, Feline, Goat, Mouse (Negative), Rat (Negative)
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.