missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vimentin Antibody (VM452), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-33060MFV500
This item is not returnable.
View return policy
Description
Vimentin Monoclonal antibody specifically detects Vimentin in Human, Porcine, Bovine, Canine, Chicken, Feline, Goat, Mouse (Negative), Rat (Negative) samples. It is validated for Flow Cytometry
Specifications
| Vimentin | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Pig, Bovine, Canine, Chicken, Feline, Goat | |
| Purified |
| Flow Cytometry | |
| VM452 | |
| Flow Cytometry | |
| epididymis secretory sperm binding protein, FLJ36605, VIM, vimentin | |
| Recombinant full-length human vimentin protein (Uniprot: P08670) | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells | |
| 7431 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction