missing translation for 'onlineSavingsMsg'
Learn More

Vimentin Antibody (RV203), DyLight 594, Novus Biologicals™

Product Code. 18429775 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429775 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429775 Supplier Novus Biologicals Supplier No. NBP197671DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Vimentin Monoclonal antibody specifically detects Vimentin in Human, Mouse, Rat, Porcine, Canine, Goat, Hamster samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vimentin
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RV203
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias epididymis secretory sperm binding protein, FLJ36605, VIM, vimentin
Host Species Mouse
Immunogen This antibody is a mouse monoclonal IgG1 antibody derived by fusion of SP2/0-Ag14 Mouse myeloma cells with spleen cells from a BALB/c Mouse immunized with a cytoskeletal vimentin extract of calf lens.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 7431
Target Species Human, Mouse, Rat, Pig, Canine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.