missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vimentin Antibody (RV202), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97672JF549
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Vimentin Monoclonal specifically detects Vimentin in Human, Mouse, Rat, Porcine, Canine, Chicken, Goat, Hamster, Primate, Xenopus, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Spezifikation
| Vimentin | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| VIM | |
| Clone RV202 is a mouse monoclonal IgG1 antibody derived by fusion of SP2/0-Ag14 mouse myeloma cells with spleen cells from a BALB/c mouse immunized with a cytoskeletal vimentin extract of calf lens. | |
| 0.1 mL | |
| Primary | |
| RV202 reacts exclusively with vimentin, which is expressed in mesenchymal cells and mesenchymal derived tumors e.g. lymphoma, sarcoma and melanoma. | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| RV202 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| FLJ36605, vimentin | |
| Mouse | |
| Protein A or G purified | |
| Biologically Active Proteins, Cancer, Cellular Markers, Cytoskeleton Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells | |
| 7431 | |
| Canine, Chicken, Goat, Hamster, Human, Mouse, Pig, Primate, Rat, Xenopus species, Zebrafish | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur