missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vimentin Antibody (RV202) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97672
This item is not returnable.
View return policy
Description
Vimentin Monoclonal antibody specifically detects Vimentin in Human, Mouse, Rat, Porcine, Canine, Chicken, Goat, Hamster, Monkey, Xenopus, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF
Specifications
| Vimentin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:1000, Flow Cytometry 1:100 - 1:200, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:100 - 1:200, CyTOF-ready | |
| epididymis secretory sperm binding protein, FLJ36605, VIM, vimentin | |
| This antibody is a mouse monoclonal IgG1 antibody derived by fusion of SP2/0-Ag14 mouse myeloma cells with spleen cells from a BALB/c mouse immunized with a cytoskeletal vimentin extract of calf lens. | |
| 0.1 mg | |
| Cancer, Cell Biology, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells | |
| 7431 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF | |
| RV202 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Canine, Chicken, Goat, Hamster, Monkey, Xenopus species, Zebrafish | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction