missing translation for 'onlineSavingsMsg'
Learn More

Vimentin Antibody, Novus Biologicals™

Product Code. 18711514 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
0.5mL
25µL
Quantity:
0.5 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize Quantity
18711514 25µL -
18738703 0.1mL -
18575892 0.5mL 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18711514 Supplier Novus Biologicals Supplier No. NB300223SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chicken Polyclonal Antibody has been used in 44 publications

Vimentin Polyclonal specifically detects Vimentin in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Host Species Chicken
Immunogen Full length recombinant human Vimentin expressed in and purified from E. coli. [Swiss-Prot# P08670]
Target Species Human, Rat, Pig, Bovine, Chicken
Form Precipitation
Isotype IgY

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.