missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vimentin Antibody (4A4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-88957
This item is not returnable.
View return policy
Description
Vimentin Monoclonal specifically detects Vimentin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Vimentin | |
| Monoclonal | |
| Unconjugated | |
| FLJ36605, vimentin | |
| Synthetic peptide corresponding to a portion of mouse vimentin phosphorylated at Ser55. | |
| 50 μL | |
| Biologically Active Proteins, Cancer, Cell Biology, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Markers, Stem Cells | |
| 7431 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, ELISA, Immunofluorescence | |
| 4A4 | |
| PBS containing 50% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction