missing translation for 'onlineSavingsMsg'
Learn More

Vimentin Antibody (2A52), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18411027 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411027 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411027 Supplier Novus Biologicals Supplier No. NBP192688MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Vimentin Monoclonal antibody specifically detects Vimentin in Human, Rat, Mouse (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vimentin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, KnockDown
Classification Monoclonal
Clone 2A52
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Knockout Validated
Formulation 50mM Sodium Borate
Gene Alias epididymis secretory sperm binding protein, FLJ36605, VIM, vimentin
Host Species Mouse
Immunogen Full length recombinant human Vimentin Antibody protein expressed in and purified from E. coli.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 7431
Target Species Human, Rat, Mouse (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.