missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Villin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116115
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Villin1.
Villin is part of cytoskeleton associated with the microvillar actin core bundle of gastrointestinal and renal brush border. It can interact with actin in a Ca2+ and phosphoinositide-regulated manner. Villin is a very specific marker for adenocarcinomas of the gastrointestinal tract and the pancreas. It is also expressed in some Merkel cell tumor and adenocarcinoma of the lung, prostate, ovarian and kidney.
Specifications
| Villin | |
| Polyclonal | |
| Unconjugated | |
| VIL1 | |
| D2S1471; OTTHUMP00000207159; Vil; Vil1; villin 1; villin-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 22349, 316521, 7429 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P09327, Q62468 | |
| VIL1 | |
| A synthesized peptide derived from human VIL1(Accession P09327), corresponding to amino acid residues D575-S625. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction