missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Villin Mouse, Clone: VIL1/1325, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £423.29
Specifications
| Antigen | Villin |
|---|---|
| Clone | VIL1/1325 |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
Specifications
| Villin | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| 7429 | |
| Recombinant human Villin fragment of 133 amino acid residues (aa179-311) | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 93kDa |
| VIL1/1325 | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| NP_009058 | |
| VIL1 | |
| IgG1 κ | |
| Protein A/G Purified | |
| Cytoplasmic and Cell Surface | |
| A549, HepG2 and HCT116 cells. Colon or Rectum. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title