missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Villin 1 Mouse anti-Human, Clone: VIL1/1314 + VIL1/2376, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07176-100ug
This item is not returnable.
View return policy
Description
Villin 1 Monoclonal specifically detects Villin 1 in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications
| Villin 1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| D2S1471, VIL, villin 1, villin-1 | |
| A recombinant fragment (around aa179-311) of human Villin 1 protein (exact sequence is proprietary) | |
| 100 μg | |
| Cancer, Cell Biology, Cellular Markers | |
| 7429 | |
| Store at 4°C. | |
| IgG1, IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| VIL1/1314 + VIL1/2376 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction