Learn More
Villin 1 Antibody (VIL1/2376), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-08928AF594
Product Code. 18514196
  • £471.00 / 100µL
Description
Villin 1 Monoclonal antibody specifically detects Villin 1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Villin 1
Monoclonal
Alexa Fluor 594
D2S1471, VIL, villin 1, villin-1
A recombinant fragment (around aa 179-311) of human Villin 1 protein (exact sequence is proprietary) (Uniprot: P09327)
100 μg
Cancer, Cell Biology, Cellular Markers
7429
Store at 4°C in the dark.
IgG1 κ
Immunohistochemistry (Paraffin)
VIL1/2376
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human
Purified