missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Villin 1 Antibody (5E3B2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52584
This item is not returnable.
View return policy
Description
Villin 1 Monoclonal specifically detects Villin 1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Villin 1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin | |
| D2S1471, VIL, villin 1, villin-1 | |
| Mouse | |
| 92.7 kDa | |
| 0.1 mg | |
| Cancer, Cytoskeleton Markers | |
| 7429 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 5E3B2 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| VIL1 | |
| Purified recombinant fragment of human VIL1 (AA: 1-209) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction