missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VILIP-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | VILIP-1 |
|---|---|
| Concentration | 0.63 mg/mL |
| Applications | Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16872344
|
Proteintech
13919-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16862344
|
Proteintech
13919-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene is a member of the visinin/recoverin subfamily of neuronal calcium sensor proteins. The encoded protein is strongly expressed in granule cells of the cerebellum where it associates with membranes in a calcium-dependent manner and modulates intracellular signaling pathways of the central nervous system by directly or indirectly regulating the activity of adenylyl cyclase. Alternatively spliced transcript variants have been observed, but their full-length nature has not been determined.Specifications
| VILIP-1 | |
| Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| P62760, P62761, P62762 | |
| 24877, 26950, 7447 | |
| VILIP-1 Fusion Protein Ag4923 | |
| Primary | |
| -20°C | |
| VSNL1 |
| 0.63 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Hippocalcin like protein 3, HLP3, HPCAL3, HUVISL1, VILIP, VILIP 1, VILIP1, visinin like 1, Visinin like protein 1, VISL1, VLP 1, VLP1, VLP-1, VSNL1 | |
| Vsnl1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title