missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vildagliptin, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
50mg
Description
Vildagliptin (LAF237) is a potent, stable, selective dipeptidyl peptidase IV (DPP-IV) inhibitor with an IC50 of 3.5 nM in human Caco-2 cells. Vildagliptin possesses excellent oral bioavailability and potent antihyperglycemic activity.
Specifications
Specifications
| Chemical Name or Material | Vildagliptin |
| CAS | 274901-16-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C17H25N3O2 |
| Quantity | 50 mg |
| Synonym | LAF237 NVP-LAF 237 |
| Solubility Information | H2O : 50 mg/mL (164.80 mM; Need ultrasonic) |
| SMILES | O[C@@]1(C2)C[C@H]3C[C@H](C[C@]2(NCC(N4[C@H](C#N)CCC4)=O)C3)C1 |
| Molecular Weight (g/mol) | 303.4 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction