missing translation for 'onlineSavingsMsg'
Learn More

Vildagliptin Carboxylic Acid Lithium Salt Hydrate, TRC

Product Code. 30453333
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
This item is not returnable. View return policy

Product Code. 30453333

Brand: Toronto Research Chemicals TRCV30501550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vildagliptin Carboxylic Acid Lithium Salt Hydrate
Analyte or Component Names Vildagliptin Carboxylic Acid Lithium Salt Hydrate
Shipping Condition Room Temperature
Molecular Formula C17H25LiN2O4
Synonym N-(3-Hydroxytricyclo[3.3.1.13,7]dec-1-yl)glycyl- L-proline Lithium Salt Hydrate
InChI Formula InChI=1S/C17H26N2O4.Li/c20-14(19-3-1-2-13(19)15(21)22)9-18-16-5-11-4-12(6-16)8-17(23,7-11)10-16;/h11-13,18,23H,1-10H2,(H,21,22);/q;+1/p-1/t11-,12+,13-,16?,17?;/m0./s1
Solution Type Neat
SMILES O=C(N1[C@H](C(O[Li])=O)CCC1)CN[C@]23C[C@](C[C@H]4C3)(O)C[C@@H](C4)C2
Industry Type Forensics
Molecular Weight (g/mol) 328.33
Quantity 50 mg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.