missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Vildagliptin anti-diabetic agent

Product Code. p-7146028 Shop All Bio Techne Products
Click to view available options
Quantity:
20 mg
100 mg
Unit Size:
100mg
20mg
This item is not returnable. View return policy

Product Code. 18235754

Brand: Novus Biologicals™ NBP26275920MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Vildagliptin is a potent inhibitor of dipeptidyl peptidase IV (DPP IV), an enzyme that cleaves glucagon-like peptide 1 (GLP-1) (1).

PubChem ID: 11077541

Application Notes: May be dissolved in DMSO (45 mg/mL); or DMF (20 mg/mL); or ethanol (20 mg/mL).

TRUSTED_SUSTAINABILITY

Specifications

CAS 274901-16-5
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Molecular Formula C17H25N3O2
Purity >98% (HPLC); NMR (conforms)
Quantity 20 mg
Source Synthetic
Target Vildagliptin anti-diabetic agent
Form Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.