missing translation for 'onlineSavingsMsg'
Learn More

Vilazodone, MedChemExpress

Product Code. 16496008
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16496008

Brand: Medchem Express HY14262

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vilazodone (EMD 68843; SB 659746A) is a potent, selective and orally active serotonin reuptake inhibitor (SSRI) and partial 5-HT1A receptor agonist. Vilazodone exhibits antidepressant efficacy in vivo can be used for the research of major depressive disorder (MDD) and affective disorders.

Vilazodone (EMD 68843

Specifications

Chemical Name or Material Vilazodone
Physical Form Solid
CAS 163521-12-8
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C26H27N5O2
For Use With (Application) Neuroscience-Neuromodulation
Synonym EMD 68843 SB659746A
Solubility Information DMSO : ≥ 100 mg/mL (226.49 mM)
SMILES NC(C1=CC2=C(C=CC(N3CCN(CCCCC4=CNC5=CC=C(C=C54)C#N)CC3)=C2)O1)=O
Molecular Weight (g/mol) 441.52
Formula Weight 441.52
Percent Purity 99.02%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.