missing translation for 'onlineSavingsMsg'
Learn More

Vilanterol trifenatate, MedChemExpress

Product Code. 16482167
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16482167

Brand: Medchem Express HY14300A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vilanterol trifenatate (GW642444 trifenatate) is a long-acting β2-adrenoceptor (β2-AR) agonist with inherent 24-hour activity. The pEC50s for β2-AR, β1-AR and β3-AR are 10.37, 6.98 and 7.36, respectively.

Vilanterol trifenatate (GW642444 trifenatate) is a long-acting β2-adrenoceptor (β2-AR) agonist with inherent 24-hour activity. The pEC50s for β2-AR, β1-AR and β3-AR are 10.37, 6.98 and 7.36, respectively.

Specifications

Chemical Name or Material Vilanterol trifenatate
CAS 503070-58-4
Color White
Purity Grade Notes Research
Molecular Formula C44H49Cl2NO7
Quantity 50 mg
Synonym GW642444 trifenatate
Solubility Information DMSO : 100 mg/mL (129.07 mM; Need ultrasonic)
SMILES OC1=CC=C([C@@H](O)CNCCCCCCOCCOCC2=C(Cl)C=CC=C2Cl)C=C1CO.O=C(O)C(C3=CC=CC=C3)(C4=CC=CC=C4)C5=CC=CC=C5
Molecular Weight (g/mol) 774.77
Formula Weight 774.77
Percent Purity 97.16%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.