missing translation for 'onlineSavingsMsg'
Learn More

Vilanterol, MedChemExpress

Product Code. 16438637
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16438637

Brand: Medchem Express HY14300

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vilanterol (GW642444) is a long-acting β2-adrenoceptor (β2-AR) agonist with 24 h activity. The pEC50s for β2-AR,β1-AR and β3-AR is 10.37±0.05, 6.98±0.03 and 7.36±0.03, respectively.

Vilanterol (GW642444) is a long-acting β2-adrenoceptor (β2-AR) agonist with 24 h activity. The pEC50s for β2-AR,β1-AR and β3-AR is 10.37±0.05, 6.98±0.03 and 7.36±0.03, respectively.

Specifications

Chemical Name or Material Vilanterol
CAS 503068-34-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C24H33Cl2NO5
Quantity 5 mg
Synonym GW642444
Solubility Information DMSO : ≥ 100 mg/mL (205.58 mM)
SMILES OC1=CC=C([C@@H](O)CNCCCCCCOCCOCC2=C(Cl)C=CC=C2Cl)C=C1CO
Molecular Weight (g/mol) 486.43
Formula Weight 486.43
Percent Purity 96.66%
Grade Research
Physical Form Oil
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.