missing translation for 'onlineSavingsMsg'
Learn More

Vilanterol Acetate, TRC

Product Code. 30450227
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
This item is not returnable. View return policy

Product Code. 30450227

Brand: Toronto Research Chemicals TRCV2600050.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vilanterol Acetate
CAS 503068-35-7
Molecular Formula C26 H37 Cl2 N O7
Quantity 0.5 mg
Synonym (α1R)-α1-[[[6-[2-[(2,6-Dichlorophenyl)methoxy]ethoxy]hexyl]amino]methyl]-4-hydroxy-1,3-benzenedimethanol Acetate (Salt), 4-[(1R)-2-[[6-[2-[(2,6-Dichlorobenzyl)oxy]ethoxy]hexyl]amino]-1-hydroxyethyl]-2-(hydroxymethyl)phenol Acetate
InChI Formula InChI=1S/C24H33Cl2NO5.C2H4O2/c25-21-6-5-7-22(26)20(21)17-32-13-12-31-11-4-2-1-3-10-27-15-24(30)18-8-9-23(29)19(14-18)16-28;1-2(3)4/h5-9,14,24,27-30H,1-4,10-13,15-17H2;1H3,(H,3,4)/t24-;/m0./s1
SMILES CC(=O)O.OCc1cc(ccc1O)[C@@H](O)CNCCCCCCOCCOCc2c(Cl)cccc2Cl
IUPAC Name acetic acid;4-[(1R)-2-[6-[2-[(2,6-dichlorophenyl)methoxy]ethoxy]hexylamino]-1-hydroxy-ethyl]-2-(hydroxymethyl)phenol
Molecular Weight (g/mol) 546.48
Formula Weight 545.1947
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.