missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VIGR/GPR126 Antibody (1044430), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne FAB10577X
This item is not returnable.
View return policy
Description
VIGR/GPR126 Monoclonal antibody specifically detects VIGR/GPR126 in Human samples. It is validated for Flow Cytometry
Specifications
| VIGR/GPR126 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| APG1, Developmentally regulated G-protein-coupled receptor, DREG, FLJ14937, G protein-coupled receptor 126, G-protein coupled receptor 126, HBV PreS1-transactivated protein 2, Vascular inducible G protein-coupled receptor, vascular-inducible G protein-coupled receptor, VIGRPS1TP2 | |
| Human embryonic kidney cell HEK293-derived human VIGR/GPR126 protein, Cys38-Lys437, Accession # AAH75798.1 | |
| 0.1 mL | |
| GPCR | |
| 57211 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry | |
| 1044430 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction