missing translation for 'onlineSavingsMsg'
Learn More

Vigabatrin, MedChemExpress

Product Code. 16422587
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16422587

Brand: Medchem Express HY15399

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vigabatrin (γ-Vinyl-GABA), an inhibitory neurotransmitter GABA vinyl-derivative, is an orally active and irreversible GABA transaminase inhibitor. Vigabatrin is an antiepileptic agent, which acts by increasing GABA levels in the brain by inhibiting the catabolism of GABA by GABA transaminase.

Vigabatrin (γ-Vinyl-GABA), an inhibitory neurotransmitter GABA vinyl-derivative, is an orally active and irreversible GABA transaminase inhibitor. Vigabatrin is an antiepileptic agent, which acts by increasing GABA levels in the brain by inhibiting the catabolism of GABA by GABA transaminase.

Specifications

Chemical Name or Material Vigabatrin
Physical Form Powder
CAS 68506-86-5
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C6H11NO2
For Use With (Application) Neuroscience-Neuromodulation
Synonym γ-Vinyl-GABA
Solubility Information H2O : 50 mg/mL (387.12 mM; Need ultrasonic)
SMILES C=CC(N)CCC(O)=O
Molecular Weight (g/mol) 129.16
Formula Weight 129.16
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.