missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ViewRNA™ Tissue Assay Core Kit

Product Code. 17152949
Change view
Click to view available options
No. of Reactions:
24 Assays
96 Assays
Unit Size:
24 tests
Pack of 96
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
17152949 96 Assays Pack of 96
16419283 24 Assays 24 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17152949 Supplier Invitrogen™ Supplier No. 19942

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The ViewRNA Tissue Assay Core Kit, when combined with a ViewRNA Tissue probe set, enables the detection of a single mRNA target in tissue sections using the Fast Red substrate.

The ViewRNA Tissue Assay Core Kit, when combined with a ViewRNA Tissue probe set (purchased separately), enables the detection of a single mRNA target in tissue sections using the Fast Red substrate. The ViewRNA Tissue Assay Blue Module enables the detection of a second mRNA target using the Fast Blue substrate and must be used in combination with the Core Kit and two probes sets in order to run a 2-plex assay. (Detect up to four RNA or microRNA in cryo or FFPE sectioned tissue with ViewRNA Tissue Fluorescence Assay, Cat. No. QVT0700) Each kit contains sufficient reagents to process a minimum of six slides at a time. ViewRNA TYPE 1 probe sets are for use with the Fast Red substrate (Core Kit), while ViewRNA TYPE 6 probe sets are for use with the Fast Blue substrate (Blue Module).

ViewRNA Tissue assays are RNA in situ hybridization (ISH) assays for the reliable detection of one or two mRNA targets within tissue sections. Visualize any gene in any tissue with single-copy sensitivity and no radioactivity. And in case your needs are not met by one of our 6500 catalog probe sets, we have decades of bioinformatics expertise and will design custom probe sets to any mRNA target of interest, free of charge.

With ViewRNA Tissue assays, you can achieve:
• Robust, single mRNA molecule detection in tissue sections
• Accuracy and specificity inherent to branched DNA amplification technologies
• Simultaneous detection of two RNA targets

The ViewRNA Tissue Assay is compatible with formalin-fixed, paraffin-embedded (FFPE) tissue. Whereas traditional ISH technologies often amplify both signal and background, ViewRNA assays use branched DNA (bDNA) technology to amplify target-specific signal and achieve superior signal-to-noise ratios.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 4 x 21 mL 100 x Pretreatment Solution (2°C to 8°C), also available separately (Cat. No. QVT0500)
• 4 x 12 mL Probe Set Diluent (2°C to 8°C), also available separately (Cat. No. QVT0510)
• 4 x 10 mL Pre-Amplifier Mix (2°C to 8°C)
• 4 x 10 mL Amplifier Mix (2°C to 8°C)
• 4 x 24 mL AP (Alkaline Phosphatase) Label Probe Diluent (2°C to 8°C), also available separately (Cat. No. QVT0513)
• 4 x 10 mL AP (Alkaline Phosphatase) Enhancer Solution (2°C to 8°C), also available separately (Cat. No. QVT0508)
• 4 x 20 mL ViewRNA Fast Red Reaction Buffer (2°C to 8°C)
• 4 x 320 μL ViewRNA Fast Red Substrate 1°C(2 to 8°C)
• 4 x 320 μL ViewRNA Fast Red Substrate 2°C(2 to 8°C)
• 4 x 320 μL ViewRNA Fast Red Substrate 3°C(2 to 8°C)
• 4 x 180 μL 100 x Protease Solution (2°C to 8°C), also available separately (Cat. No. QVT0512)
• 4 x 45 μL AP (Alkaline Phosphatase) Label Probe Type 1°C(2 to 8°C)
• 4 x 144 mL 100 x Wash Buffer Component 1°C(15 to 30°C)
• 4 x 280 mL 400 x Wash Buffer Component 2°C(15 to 30°C)

Format Kit
No. of Reactions 96 Assays
Product Line ViewRNA
Product Type RNA In Situ Hybridization Tissue Assay Core Kit
Quantity 96 Assays
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.