missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ViewRNA™ Tissue Assay Blue Module

Product Code. 16395142
Change view
Click to view available options
No. of Reactions:
24 Assays
96 Assays
Unit Size:
24 tests
Pack of 96
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
16395142 24 Assays 24 tests
17162949 96 Assays Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16395142 Supplier Invitrogen™ Supplier No. 19932

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This VewRNA Tissue Assay Blue Module is designed for use with the ViewRNA Tissue Assay Core Kit.

This VewRNA Tissue Assay Blue Module is designed for use with the ViewRNA Tissue Assay Core Kit. The Core Kit, when combined with a ViewRNA Tissue probe set (purchased separately), enables the detection of a single mRNA target in tissue sections using the Fast Red substrate. The ViewRNA Tissue Assay Blue Module enables the detection of a second mRNA target using the Fast Blue substrate and must be used in combination with the Core Kit and two probes sets in order to run a 2-plex assay. (Detect up to four RNA or microRNA in cryo or FFPE sectioned tissue with ViewRNA Tissue Fluorescence Assay, Cat. No. QVT0700.) Each kit contains sufficient reagents to process a minimum of six slides at a time. ViewRNA TYPE 1 probe sets are for use with the Fast Red substrate (Core Kit), while ViewRNA TYPE 6 probe sets are for use with the Fast Blue substrate (Blue Module).

ViewRNA Tissue assays are RNA in situ hybridization (ISH) assays for the reliable detection of one or two mRNA targets within tissue sections. Visualize any gene in any tissue with single-copy sensitivity and no radioactivity. And in case your needs are not met by one of our 6500 catalog probe sets, we have decades of bioinformatics expertise and will design custom probe sets to any mRNA target of interest, free of charge.

With ViewRNA Tissue assays, you can achieve:
• Robust, single mRNA molecule detection in tissue sections
• Accuracy and specificity inherent to branched DNA amplification technologies
• Simultaneous detection of two RNA targets

The ViewRNA Tissue Assay is compatible with formalin-fixed, paraffin-embedded (FFPE) tissue. Whereas traditional ISH technologies often amplify both signal and background, ViewRNA assays use branched DNA (bDNA) technology to amplify target-specific signal and achieve superior signal-to-noise ratios.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 45 μL Label Probe 6-AP (2°C to 8°C)
• 420 μL Blue Reagent 1°C(2 to 8°C)
• 420 μL Blue Reagent 2°C(2 to 8°C)
• 420 μL Blue Reagent 3°C(2 to 8°C)
• 20 mL Blue Buffer (2°C to 8°C)
• 10 mL AP Stop QT (2°C to 8°C)

No. of Reactions 24 Assays
Product Line ViewRNA
Product Type RNA Tissue Assay Blue Module
Quantity 24 Assays
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.